Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Membrin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-PT046-F VAM-PT046-D
89133-292EA 576 USD InStock
89133-292 89133-290
Anti-Membrin Mouse Monoclonal Antibody
Antibodies
Membrin, also known as GS27 (Golgi SNARE of 27K), is an integral membrane protein present on the surface of the Golgi apparatus. GS27 is a member of a protein complex that consists of cis-Golgi vesicle receptor syntaxin 5 and GS28, v-SNARE rbet1 and Rsec22, and Rsly1. These complexes are postulated to direct the fusion of ER-derived vesicles with vesicular tubular cluster (VTC), and the fusion of VTCs to form cis-Golgi compartment.

  • For Immunocytochemistry, Western Blot

Recognizes human, rat, dog, chicken, hamster, monkey, pig and rabbit Membrin. Detects a band of ~27kDa by Western blot.

Type: Primary
Antigen: MEMBRIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty