Would you like to log in again?
Testosterone High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-176 ADI-901-176
89133-300EA 1890 USD InStock
89133-300 89133-298
Testosterone High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
A highly sensitive competitive immunoassay for the measurement of Testosterone in human serum and plasma samples
  • Excellent sensitivity
  • Validated for use with human samples
  • Stable color-coded reagents reduce errors
For the quantitative determination of human testosterone in plasma, serum, and urine samples.
Results are read on a colorimetric plate reader at 405 nm.
Ordering information: Alternative size available as ADI-900-176 (1x96 wells)
Delivery information: Kit includes GxM microtiter plate, assay buffer, standard, antibody, conjugate, wash buffer concentrate, pNpp substrate, stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty