Would you like to log in again?
Serotonin ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-175
89133-388EA 756 USD InStock
89133-388
Serotonin ELISA Kit, Enzo Life Sciences
Assays ELISAs
The Serotonin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours.
  • Highly sensitive measurement of serotonin
  • No acylation required for standards and samples
  • Validated in multiple complex sample matrices
  • High throughput format with results
  • Fully quantitative results that surpass semi-quantitiave Western blot analysis
For the quantitative determination of Serotonin in platelets, plasma, serum, and urine from any species.
Delivery information: Supplied with GxR IgG microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty