Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDI Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-891-F ADI-SPA-891-J ADI-SPA-891-D SPA-891J
89133-404EA 590 USD InStock
89133-404 95046-300 89133-402 CA95046-300
Anti-PDI Mouse Monoclonal Antibody
Antibodies
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot, EM

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: PDI
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty