Would you like to log in again?
Urea Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
MM111-500G MM111-500G
89133-744EA 118.97 USD InStock
CA89133-744 89133-744
Urea Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the identification of bacteria on the basis of urea utilization.
  • Identify bacteria species differentiation
  • Light yellow to light pink colored
  • Homogeneous, free flowing powder
For the identification of bacteria on the basis of urea utilization, specifically for the differentiation of Proteus species from Salmonella and Shigella species in accordance with Indian Pharmacopoeia, 2007.
Caution: Store below 8 °C in tightly closed container and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty