Would you like to log in again?
Amplitube™ PCR Reaction Strips with Attached Caps, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitube™
T320-3G T320-2B T320-2R T320-2B T320-2G T320-3Y T320-3N T320-2N T320-2N T320-3B T320-3R T320-3B T320-3R T320-2R T320-3Y T320-2Y T320-3G T320-2G T320-2Y T320-3N
89133-914PK 165.85 USD InStock
CA48648-642 CA11028-440 CA11028-450 CA48648-640 CA11028-444 CA11028-442 89133-914 89133-924 89133-912 89133-922 89133-910 89133-920 89133-918 89133-928 89133-916 89133-926 CA11028-448 CA11028-436 CA11028-446 CA11028-438
Amplitube™ PCR Reaction Strips with Attached Caps, Simport Scientific
Tubes PCR Tubes
This 0.2 ml tube strip incorporates individually attached caps, eliminating the need to carry two separate components in inventory.
  • Autoclavable
This PCR reaction strip is available with either flat or dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the entire thermal cycle. The cap has an integral seal preventing contamination with the lid surface.
Designed for easier handling than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. They are manufactured under the most stringent conditions to attain the highest quality industry standards. Available in a choice of colorless or four non-cytotoxic and non-metallic color varieties.
The strip includes 8 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer.
Certifications: Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Packaging: Packed in tamper-proof resealable bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty