Would you like to log in again?
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B16-100 100B B26-150 100B B22-150 B18-100 B26-100 B18-50 100BK B18-50 B25-100 100B B24-100 1000 B21-100 100B B19-100 100B B22-50 100BK B27-100 100B B20-150 100B B22-100 1000 B30-100 1000 B19-50 100BK B26-50 100BK B22-100 B21-150 B21-100 B19-150 100B B24-50 100BK B23-100 1000 B18-100 B23-150 100B B26-150 B27-100 B24-50 B16-100 B23-100 100B B19-150 1000 B23-50 B19-50 1000B B24-150 100B B19-150 B26-100 100B B27-150 B20-150 B21-100 1000 B18-50 1000B B25-50 B30-100 100B B25-150 B20-50 100BK B19-100 B21-50 1000B B25-100 B20-100 1000 B22-50 1000B B20-150 1000 B24-150 B22-100 100B B23-50 100BK B15-100 100B B22-50 B30-100 B22-150 100B B18-100 100B B21-150 100B B18-150 100B B21-50 100BK B21-150 1000 B20-50 B25-100 1000 B26-150 1000 B18-150 B25-150 100B B21-50 B27-50 100BK B18-100 100B B24-150 1000 B25-150 1000 B27-150 100B B30-50 1000B B27-50 1000B B27-50 B22-150 1000 B19-50 B23-50 1000B B24-50 1000B B20-100 100B B20-50 1000B B26-50 1000B B16-100 1000 B23-150 B27-100 1000 B30-50 B26-50 B26-100 1000 B24-100 100B B19-100 1000 B15-100 B23-100 B25-50 1000B B25-50 100BK B18-150 1000 B15-100 1000 B30-50 100BK B20-100 B23-150 1000 B27-150 1000 B18-100 1000 B18-100 1000 B24-100
89134-092PK 44.16 USD InStock
89134-092 89134-094 89134-090 89134-012 89134-132 89134-014 89134-134 89134-096 89134-010 89134-098 89134-130 89134-016 89134-136 89134-018 89134-138 89134-080 89134-082 89134-122 89134-088 89134-124 89134-084 89134-120 89134-086 89134-008 89134-126 89134-128 89134-034 89134-154 89134-036 89134-156 89134-030 89134-150 89134-032 89134-152 89134-038 89134-158 89134-144 89166-250 89134-022 89166-252 89134-146 89134-024 89134-140 89166-254 89134-142 89166-256 89134-020 89134-148 89134-026 89134-028 89134-050 89134-170 89134-056 89134-176 89166-240 89134-058 89134-178 89166-242 89134-052 89134-172 89166-244 89134-054 89166-246 89134-174 89166-248 89134-160 89134-166 89134-044 89134-168 89134-046 89134-162 89134-040 89134-164 89134-042 89134-048 89134-070 89134-190 89134-072 89134-192 89134-078 89134-198 89134-110 89134-112 89134-074 89134-194 89134-076 89134-196 89134-118 89134-114 89134-116 89134-180 89134-182 89134-060 89134-100 89134-188 89134-066 89134-102 89134-068 89134-184 89134-062 89134-186 89134-064 89134-108 89134-104 89134-106
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Cannulae
These Luer Stub Adaptors (Blunt Needles) offer superior quality at an affordable price. With a plastic Luer hub and stainless-steel cannula, these blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters and dispensing of liquids. The smooth and burr-free stainless-steel needles will not damage the inner lumen when accessing catheters or tubing. Ensure the integrity of the catheter when dosing, sampling, flushing, and locking with these blunt needles.
  • Sterile needles are EtO exposed and individually packaged (25 per pack)
  • Non sterile needles are affordably packaged in bulk quantities of 100 or 1000
  • High-quality, low cost
  • Stainless steel cannula has a burr-free tip
  • Also available attached to empty or pre-filled flushing or locking syringes
Blunt Tip Needles (Luer Stub Adaptors) have countless laboratory applications. For instance, when dosing, sampling, or infusing, these blunt needles ensure a secure and reliable connection to in-dwelling catheters. The superior manufactured blunt needles provide a smooth, burr-free needle edge which prevents inadvertent damage to the lumen of the catheter caused by inferior needles. These high-quality, low-cost blunt needles are a critical tool for dosing, sampling, flushing, and locking catheters.
Ordering information: Contact a Avantor representative for more information on additional options.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty