Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isotonic Saline (NaCl) Prefilled Syringes, SAI Infusion Technologies

Supplier: SAI Infusion Technologies
SC1-1.0
89134-214CS 248.6 USD InStock
89134-214
Isotonic Saline (NaCl) Prefilled Syringes, SAI Infusion Technologies
Syringes Syringes, Medical
Prefilled syringes for flushing an indwelling catheter or port system. This prefilled option can saves time, space, and money.

  • Saline (0.9%) filled syringe, customization available
  • Syringe sizes from 1 to 60 ml
  • Volumes from 0.2 ml to filled
  • Syringes with caps, or with blunt needles already attached

No need to unwrap individual syringes, no need to store physiologic containers, no need to formulate solutions, and no need to spend valuable time filling syringes.

Sterile solution and fluid path. Exterior surface of syringe is non sterile.

Heparinized saline syringes and catheter maintenance kits are also available.

Ordering information: Minimum order amount of 3 boxes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty