Would you like to log in again?
VWR® Sealing Film, Polyester
Supplier: Avantor
VWR®
THNL100 THSNL100
89134-430CS 101.38 USD InStock
89134-430 89134-432
VWR® Sealing Film, Polyester
Seals Microplate Seals
Polyester film, acrylic adhesive. These 49.5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
  • Recommended temperature range from −40 to +120 °C
  • Available sterile and non sterile
  • Marginally pierceable by single channel pipette
  • Applications: Short-term storage, incubation, ELISA
Each film L×D: 141,0×79,4 mm
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty