Would you like to log in again?
VWR® Adhesive PCR Film Seals
Supplier: VWR
VWR®
TSSVWR100 TSANL100 TSVWR100 TS2X8NL50
60941-072CS 164.61 USD InStock
60941-072 60941-070 89087-690 89134-434
VWR® Adhesive PCR Film Seals
Seals Microplate Seals
These heat resistant 74.9 µm thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For Real-Time PCR applications, where maximum optical clarity is required, use optically-clear polyester films.
  • Recommended for temperatures from −40 to +120 °C
  • Certified free from DNase, RNase and nuclease
Each film L×D: 135,1×79,4 mm (55/16x31/8") with sufficient sealing area for all PCR plates.
Length with end tabs removed: 123,1 mm (47/8")
* stronger, thicker adhesive and cut to fit raised-rim plates
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty