Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Type I Safety Cans for various types of liquids, Justrite®
Supplier: Justrite
7120210 7150200 7125210 7125300 7120200 7120410 7150410 7110400 7150300 7120400 7125310 7125200 7150310 7110210 7125400 7120300 7110200 7125410 7120310 7110310 7110410 7150210 7150400 7110300
89134-606EA 160.6 USD InStock
89134-606 89134-608 89134-562 89134-584 89134-572 89134-550 89134-564 89134-586 89134-596 89134-552 89134-580 89134-582 89134-570 89134-548 89134-604 89134-610 89134-566 89134-588 89134-554 89134-598 89134-546 89134-568 89134-578 89134-556
Type I Safety Cans for various types of liquids, Justrite®
Safety Cans
Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer
Blue- kerosene, Yellow - diesel, Green-oils.
Protection features include a 3-1/2" (88mm) long chemical resistant stainless steel flame arrester that stops flashback ignition, provides faster liquid flow, and exceptional durability. The self-venting, self-closing leakproof lid prevents ruptures from pressure build-up and controls vapors and spills. Warning label includes an extra large ID zone for user labeling of contents to avoid misuse, reduce the potential for incompatible mixtures, or may be used to designate department or individual owner. Made of galvanized steel with a tough powder paint finish, can capacities 3.8L (1gal.) and above have ample headspace to accommodate oil additives.
Certifications: FM. UL, ULC, TUV.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty