Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EcoPolyBlend™ Spill Containment Caddies, Justrite®

Supplier: Justrite
28665
89134-668EA 1416.96 USD InStock
89134-668
EcoPolyBlend™ Spill Containment Caddies, Justrite®
Drum Storage
These caddies perform three key chemical handling functions: safe drum transport, mobile dispensing, and spill containment.

  • Perform three key chemical handling functions: safe drum transport, mobile dispensing, and spill containment
  • Made from durable, 100% recycled polyethylene for long life and chemical resistance
  • Designed to minimize back injuries and muscle strain
  • Safely dispense flammable liquids in horizontal position
  • Unique, open-view sump for easy spill detection and easy clean up
  • Nylon strap secures 100 or 200L (30 or 55gal.) drum in place

Made from durable, 100% recycled polyethylene for long life and chemical resistance, designed to minimize back injuries and muscle strain. Safely dispense flammable liquids in horizontal position. Unique, open-view sump for easy spill detection and easy clean-up. Nylon strap secures 100 or 200L (30 or 55gal.) drum in place. Outdoor caddy features rubber wheels to absorb bumps.

Color: black

Certifications: Meet EPA 40 CFR 264.175, NFPA, and SPCC requirements

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty