Would you like to log in again?
EcoPolyBlend DrumSheds™, Justrite®
Supplier: Justrite
28676 28675 28679
89134-694EA 700.97 USD InStock
89134-694 89134-686 89134-688
EcoPolyBlend DrumSheds™, Justrite®
Drum Storage
Eco-friendly, weather-resistant drum storage.
  • Spacious interior for funnels or pumps
  • Leak-proof pallet sump meets EPA and SPCC requirements
  • Forklift pockets front and back for easy transport
  • Ships fully assembled
Weather-resistant outdoor sheds offer high performance spill control. A robust design made of 55% recycled polyethylene features convenient rolltop doors to allow access from front or rear. Forklift pockets on the front and back enable easy transport and relocations.The EPA/SPCC compliant leakproof pallet sump features thick, durable deck grating that can be removed for easy inspection and clean up of spills.Wide door opening of 132.1cm(52”) facilitates easy storage palletized loads. A spacious interior offers ample headspace to accommodate working operations with funnels or pumps. Safety features include grounding capability and integral anchor bolt locations for wind resistance. Molded in lugs accept optional padlocks for security. Ships fully assembled to save on set-up time. Optional ramp simplifies drum placement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty