Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FILTRA-BAG® Blender Bags, Labplas
Supplier: Labplas
FILTRA-BAG®
SCT-1520A EFT-1015A SCT-1520A
89134-808CS 374.09 USD InStock
CA89134-808 89134-808 89182-944
FILTRA-BAG® Blender Bags, Labplas
Bags Blender Bags
FILTRA-BAG ® blender bags are designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid / solid substances.
When lacing an aliquot onto growth media, it is very important that the volume of the aliquot be accurate and that it be free of solid particles. This prevents pipette blockage as well as uneven plate distribution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty