Would you like to log in again?
MegaSwat™ Mops, Micronova
Supplier: Micronova
MegaSwat™
QDMS-8 MS1111-16 MS1111-8 MS6-8IR MS1111-16IR MS1111-8IR MS6-8 MSAA-16NS MS6-16NS MSAW-24NS MSAA-8NS QDMS-16
76323-708CS 333.1 USD InStock
76323-708 10142-782 89508-390 89134-960 76323-706 89134-958 76323-712 68400-208 76323-710 76292-768 89092-750 68400-412
MegaSwat™ Mops, Micronova
Mops
The MegaSwat™ blade-style mop is ideal for cleaning around, under, or behind heavy process equipment or calibrated apparatus in the lab and cleanroom. Featuring a flexible polyethylene blade with a choice of covers. Lightweight and easy to use, the low-profile mop can be used for wet or dry cleaning or disinfecting in ISO 5 - 9 environments.
  • Single-ply and foam lined mops easily glide under and behind equipment
  • Lengths range from 8 to 36", and widths range from ⁵/₈ to 2"
  • Compatible with alcohol, strong disinfectants, and steam sterilization
  • Autoclavable covers and hardware
  • Launderable covers
The MegaSwat™ mop adapter is available in a range of lengths and widths. The adapter is designed primarily to work with the SSU series QD handles (there is a QP version for threaded handles). Corresponding MegaSwat™ mops are available with or without a foam interior and gamma irradiated for aseptic applications. The range of fabrics to match cleaning needs include: NovaPoly 100% polyester knit, which provides excellent disinfectant dispersal. PolySorb 100% textured polyester, which assists in removing residues and dispersing disinfectant. MegaTex 100% non-woven polyamide, offering slight abrasion with good absorption. Microfiber, 80% polyester/20% polyamide, absorbent wet or dry.
Ordering information: Mop cover, adapter, and handle are ordered separately. For additional handle lengths, see 68400-302 series.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty