Would you like to log in again?
Luria Bertani Agar, Miller, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1151-10KG M1151-10KG M1151-500G M1151-500G M1151-5KG M1151-2.5KG M1151-2.5KG M1151-5KG
89135-064EA 1310 USD InStock
89135-064 CA89135-064 95020-584 89223-128 CA89223-128 CA95020-584 CA89129-482 89129-482
Luria Bertani Agar, Miller, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and maintenance of recombinant strains of Escherichia coli for genetic and molecular studies.
  • Microbiological culture media
  • 500 g poly bottles
Suitable for routine isolation cultivation of microorganisms that are not particularly fastidious.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty