Would you like to log in again?
MOP ULTRAMOP ST.81X5X16IN CS50
Supplier: FOAMTEC INTERNATIONAL, LLC.
Sterile Sponge Mop Head^ .81X5X16In Clean Room Sterile Foam Mop Head Designed To Fit The Truclean Mopping System. Foam Mop Refills Are Constructed Of Open Cell Polyurethane Foam That Cleans, Absorbs Spills And Applies Even Volumes Of Disinfectants. Individually Packed To Maintain Cleanliness. Gamma Irradiated To Sal 10-6. 50 Per Case
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MOP ULTRAMOP ST.81X5X16IN CS50 89135-140 FS500ST Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty