Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP65 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-881-F ADI-SPA-881-D
89135-226EA 560 USD InStock
89135-226 89135-224
Anti-HSP65 Mouse Monoclonal Antibody
Antibodies
Hsp65 is a member of the Hsp60 family of heat shock proteins isolated from Mycobacterium bovis BCG. Hsp65 from M. bovis is identical to that of M. tuberculosis, and similar to that of M. leprae, two highly pathogenic strains of mycobacterium. Hsp65 is the immunodominant antigen during mycobacterial infection and vaccination, and has been linked to the development of autoimmune adjuvant arthritis in rats.

  • For Western Blot

Recognizes human, mouse, rat, bovine, bacteria (E. coli, M. leprae, M. bovis), cryptosporidium, Drosophila, hamster and guinea pig HSP65. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: HSP65
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty