Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NF-kappa B P65 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAS-TF110-F KAS-TF110-D
89135-314EA 233 USD InStock
89135-314 89135-316
Anti-NF-kappa B P65 Rabbit Polyclonal Antibody
Antibodies
Nuclear factor kappa-B (NFk) B) functions as a sequence specific transcriptional activator that binds to the intronic enhancer of kappa light chain gene in B lymphocytes. NF-kB is a heterodimer consisting of a 50 kDa DNA binding subunit (p50) and a 65 kDa transactivation subunit (p65/RelA) (ref. 3), both of which exhibit sequence homology to the protooncogene c-Rel. p50 has an isoform called p49/p52, and both proteins derive from the amino-terminal of precursor protein p105 and p100, respectively. The p50/p65 heterodimer remains in inactive form in the cytosol as a complex with its inhibitor, IkB. Upon stimulation of cells by a variety of stimuli, including lipopolysaccharide (LPS), pro-inflammatory cytokines like interleukin-1 and tumor necrosis factor, and viral infection, IkB becomes phosphorylated and degraded by the proteosome. The active NF-kB heterodimer translocates into the nucleus and induces gene expression.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, monkey, pig and rabbit NF-kappaB p65. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: NF-KAPPAB P65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty