Would you like to log in again?
[pSer326]HSF1 ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-199
89135-346EA 674 USD InStock
89135-346
[pSer326]HSF1 ELISA kit, Enzo Life Sciences
Assays ELISAs
The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 phosphorylated at Ser326 in cell lysates and tissue extracts from human and mouse origins
  • Use less precious sample compared to Western blot
  • Time saving and convenient
  • Ready to use liquid color coded reagents and pre-coated plate
For the quantitative determination of human and mouse [pSer326]HSF1 in cell lysates and tissue.
This is a sensitive ELISA kit for the key transcription factor for heat shock protein (chaperone) research.
Delivery information: Kit includes antibody, assay buffer, conjugate, extraction reagent #1, microtiter plate, standard, stop solution, TMB substrate, wash buffer concentrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty