Would you like to log in again?
Corning® DeckWorks™ Barrier Pipette Tips, Hinged Rack, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
4140 4138 4137 4136 4135 4139
89136-161CS 609.66 USD InStock
89136-161 89136-163 89136-155 89136-165 89136-157 89136-159
Corning® DeckWorks™ Barrier Pipette Tips, Hinged Rack, Corning
Pipette Tips
These low binding barrier tips feature an inert, high-density PE barrier to eliminate aerosol carryover contamination during critical pipetting procedures. The tips will not trap liquids or inhibit PCR in the event of over pipetting.
  • Tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes
  • Non pyrogenic
  • Human DNA-free, RNase-/DNase-free
  • Sterility Assurance Level 10⁻³
This system is designed to help reduce the production and waste of plastic materials. The hinged racks are manufactured with recycled plastic.
Corning DeckWorks barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks tips will not trap liquids or inhibit PCR in the event of over-pipetting.
Corning DeckWorks proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. All tips are manufactured from polypropylene.
Ordering information: Corning DeckWorks pipette tips are also available in bulk, hinged rack, reload, reload starter kit, and tip station configurations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty