Would you like to log in again?
Corning® DeckWorks™ Pipette Tips, Tip Station, Corning
Supplier: Corning
Corning® DeckWorks™
4144 4141 4143 4142
89136-171CS 405.31 USD InStock
89136-171 89136-173 89136-167 89136-169
Corning® DeckWorks™ Pipette Tips, Tip Station, Corning
Pipette Tips
These tip stations maximize bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
  • Ideal single and mult-ichannel pipetting stations
  • Five-layer deck design (2×96) matches the microtiter plate format
  • Stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities. These tip stations are offered both sterile and non sterile.
Certified Free of: RNase, DNase, Pyrogens, PCR Inhibitors.
Both the hinged racks and relaod decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimize the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tip s are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Ordering information: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload and reload starter kit configurations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty