Would you like to log in again?
VWR® Basic SPP Lab Coats
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-349 89208-812 89208-814 414004-342 89208-750 414004-350 89208-756 89208-754 89208-816 414004-354 414004-348 414004-343 89136-824 414004-353 414004-344 89208-818 89136-828 89208-752 414004-352 414004-345 89208-820 414004-351 414004-346
89208-812CS 159.9 USD InStock
89208-812 414004-348 89208-756 89208-814 89136-824 414004-346 414004-345 414004-344 414004-354 414004-343 89208-752 414004-342 414004-353 89136-828 89208-754 414004-352 89208-820 89208-816 89208-818 414004-349 414004-351 414004-350 89208-750
VWR® Basic SPP Lab Coats
Disposable Garments
VWR® Basic Protection SPP Lab Coats are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable
  • Economical, disposable first-line barrier
  • Maximum breathability
  • Low linting
Garments feature inset sleeves, snap front, and tapered collar. Available with elastic cuffs or knit cuffs. Serged seams. Lab coats are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They may be used to protect against dirt, grime, and certain dry particulates in nonhazardous environments. These knee-length, general-purpose garments are ideal for less critical areas or pre-gowning entry rooms.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty