Would you like to log in again?
Laboratory Bottles, Narrow Mouth, with Thread, BrandTech
Supplier: BrandTech
704006 704008 704004
89136-836EA 65.99 USD InStock
89136-836 89136-834 89136-832
Laboratory Bottles, Narrow Mouth, with Thread, BrandTech
Bottles Glass Bottles
Soda-lime glass (amber).
  • Suitable for Dispensette®, seripettor® and other bottletop dispensers
  • Amber soda-lime glass
  • Pouring ring and screw cap included
The ethylene-acrylate coating envelops the glass bottle like a protective skin. Should the bottle break, the splintering hazards are reduced considerably. The maximum operating temperature for coated bottles is 80 °C.
Note: To preserve the coating, do not clean at temperatures exceeding 60 °C.
Screw cap PP, pouring ring PE-LD. Space-saving square base. The bottles are available ethylene-acrylate coated and uncoated.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty