Would you like to log in again?
Advanced TC™ Treated 96-Well Cell Culture Microplate, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
655980 655986 655983 675983 675986 655982
89131-694CS 375.24 USD InStock
89131-694 89136-852 89131-692 89136-850 89136-856 89136-854
Advanced TC™ Treated 96-Well Cell Culture Microplate, Polystyrene, Sterile, Greiner Bio-One
Microplates Cell Culture Plates
Advanced Tissue Culture (TC) treated 96 Well Microplates, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Sterile
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Noncytotoxic
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield.
All plates come with lid.
Ordering information: Lid included with Condensation Rings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty