Would you like to log in again?
Lab-Tips® Long Open Cell Foam Swab, Berkshire
Supplier: Berkshire
Lab-Tips®
LT01465.10
89137-168CS 307.59 USD InStock
89137-168
Lab-Tips® Long Open Cell Foam Swab, Berkshire
Swabs Swabs for Cleaning
Berkshire's Lab-Tips® swabs are exceptionally pure for precision cleaning, surface sampling, applying lubricants and other liquids, and validation procedures.
  • Low particles, fibers, ions, and extractables
  • Good chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Good sorbency
  • Free from silicone, amides, and DOP
Reticulated or open cell foams are structures in which the cell windows have been removed by thermal or chemical means. This process results in a structure that has excellent liquid retention and particle entrapment capability. Berkshire's open cell foam swabs are made from clean room laundered 100 ppi polyurethane foam for superior cleanliness.
Lab-Tips® are double bagged in bags manufactured from high purity polyethylene resin that ensures ultra-low organic and inorganic residues. Bags are free from silicone, amides, and DP and comply with RoHS directives. An easy tearnotch and solvent resistant inner pack labels ensure ease of use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty