Would you like to log in again?
1536-Well Non-binding Microplate, Nonsterile, Greiner Bio-One
Supplier: Greiner Bio-One
782900 782904
89137-768CS 1578.75 USD InStock
89137-768 89137-770
1536-Well Non-binding Microplate, Nonsterile, Greiner Bio-One
Microplates Deep Well Microplates
Greiner Bio-One's non-binding microplate surface is characterized by low protein, DNA and peptide binding properties which improve data quality especially in homogeneous methods such as Fluorescence Polarization Assays (FPA) or Scintillation Proximity Assays (SPA).
  • Nonsterile
  • Available with solid or clear film well bottoms
  • Non-binding surface
  • No potential for degradation or leaching
This non-binding surface is stable under common assay conditions, there is no potential for degradation or leaching and resultant assay interference. Non-binding microplates are featured in 96, 384 and 1536 well formats in black, white, and clear, including solid and µClear film well bottoms. Also available in SV Hibase design.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty