Would you like to log in again?
cAMP and cGMP conjugates, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
00022-1 00021 00021-1 00020 00022 00020-1
89138-106EA 505.92 USD InStock
89138-106 89138-104 89138-102 89138-110 89138-100 89138-108
cAMP and cGMP conjugates, Biotium
Stains Histology Stains
Conjugates for probing cAMP receptors or measuring cGMP in immunoassays.
  • Biotin-cAMP can be used to probe cAMP receptors
  • Biotin-cGMP has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples
  • Cyanine 644-cAMP can be used to probe cAMP receptors
Cyanine 644-cAMP λEx/λEm (MeOH) = 644/670 nm
Biotin-cAMP, diisopropylethylammonium salt is a biotin-conjugated cAMP analog that can be used to probe cAMP receptors. Biotin-cGMP, diisopropylethylammonium salt has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples. Cyanine 644-cAMP is a fluorescently labeled cAMP analog and can be used to probe cAMP receptors.
Single molecules of fluorescently labeled cAMP have been imaged as the probe binds to the receptor on the surface of Dictyostelium cells.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty