Would you like to log in again?
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Supplier: Biotium
10013 10019 10024 10018 10022 10003 10020 10015 10006 10007 10004 10030 10014 10005 10012 10017 10025
89138-248EA 261.06 USD InStock
89138-248 89138-226 89138-238 89138-224 89138-236 89138-200 89138-222 89138-244 89138-234 89138-242 89138-198 89138-232 89138-240 89138-196 89138-194 89138-192 89138-228
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Protein Linking and Labeling Systems
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
  • Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
  • Chromogenic substrates for absorbance-based detection or light microscopy
  • Fluorogenic substrates for highly sensitive fluorescence-based ELISA
  • Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty