Would you like to log in again?
Coelenterazine Substrates for Renilla Luciferase and Apoaequorin, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10117-2 10116-2 10115-1 10126 10110-1 10126-50UG 10125-1 10125-2 10125 10113-2 10115-2 10113-1 10114-2 10122-1 10111-1 10127 10113 10116-1 10121-1 10110-2 10124-2 10111 10116 10121 10121-2 10112-1 10122 10124 10127-50UG 10111-2 10115 10117 10112-2 10114 10123 10117-1 10110 10114-1 10112
89410-672EA 178.77 USD InStock
89410-672 89138-326 89138-304 89138-348 89138-324 89138-346 89138-302 89138-322 89138-344 89138-300 89138-342 89138-320 89138-340 89138-318 89138-338 89138-316 89138-336 89138-314 89138-312 89138-334 89138-310 89138-332 89138-330 89138-352 89138-298 89138-350 75832-548 89138-296 89138-294 89138-292 75832-546 89138-290 89411-152 89493-038 89493-036 89411-156 89138-308 89138-328 89138-306
Coelenterazine Substrates for Renilla Luciferase and Apoaequorin, Biotium
Protein Linking and Labeling Systems
Coelenterazine and its analogs are luminescent enzyme substrates for apoaequorin and Renilla luciferase.
Coelenterazine and its analogs are luminescent enzyme substrates for apoaequorin and Renilla luciferase. Apoaequorin is used as a calcium indicator whereas Renilla luciferase is commonly used as a reporter of transcription regulation. Coelenterazine binds to apoaequorin to form aequorin which emits light upon binding to calcium. Aequorin can be used to measure a broad concentration range of calcium from ~0.1 uM to >100 uM. Renilla luciferase has been widely used as a reporter protein and as a bioluminescence donor in bioluminescence resonance energy transfer (BRET) to study protein-protein interactions. Other uses of coelenterazine include chemiluminescent detection of superoxide anion and peroxynitrite in cells or tissues.
Can’t decide which coelenterazine analog to use? Try the Coelenterazine Sampler Kit (catalog no. 89138-348). The kit includes 25 ug each of nine coelenterazine analogs including native, cp, f, fcp, h, hcp, i, ip and n.
There are over a dozen of coelenterazine analogs that have been synthesized, many of which are now commercially available from Biotium. These coelenterazine analogs have different properties in terms of emission wavelength, cell membrane permeability and quantum efficiency.
Biotium offers coelenterazine 400a (also known as DeepBlue C™) (catalog no. 89138-350) which serves as a substrate for Renilla luciferase (Rluc) and generates an emission peak centered around 400 nm. It is the preferred Renilla luciferase substrate for BRET studies because it has minimal interference with the emission of the GFP acceptor.
Also, see Biotium's water soluble Aquaphile™ Coelenterazines, sold as coelenterazine native (catalog no. 89411-152) or coelenterzine h analog (catalog no. 89411-156). These formulations readily dissolve in water or buffer for in vivo use.
Lastly, Biotium offers methyl coelenterazine (coelenterazine, 2-methyl analog) (catalog no. 89138-344) which has been reported to be a superior antioxidant for cells against reactive oxygen species (ROS) such as singlet oxygen and superoxide anion. The coelenterazine derivative is membrane-permeant, non-toxic and highly reactive toward ROS.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty