Would you like to log in again?
Annexin V Near IR CF® Dye Conjugates, Biotium
Supplier: Biotium
29007 29006 29046 29082 29070 29078 29047
76221-384EA 375.42 USD InStock
76221-384 89138-862 89138-860 75832-834 76221-390 89427-072 10018-152
Annexin V Near IR CF® Dye Conjugates, Biotium
Protein Linking and Labeling Systems
Near-IR Annexin V fluorescent conjugates can be used to detect apoptotic cells and is suitable for in vivo imaging.
  • Detect apoptotic cells with fast, reliable detection of phosphatidylserine externalization
  • No azide or other potentially toxic materials or preservatives, suitable for in vivo imaging
  • Choice of 7 near-IR fluorescent CF® dye colors
  • Biotium's CF® dyes are far more photostable and brighter than other similar near-IR dyes
  • Can be used for fluorescence microscopy, live cell imaging, and flow cytometry
Annexin V is a 35 to 36 kDa protein that has a high affinity for phosphatidylserine (PS). In normal viable cells, PS is located on the cytoplasmic surface of the cell membrane. In apoptotic cells, however, PS is translocated from the inner to the outer leaflet of the plasmic membrane, making itself available for Annexin V binding. Near-IR CF® Dye Annexin V does not contain azide or other potentially toxic materials, and thus is suitable for in vivo imaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty