Would you like to log in again?
EvaEZ™, Fluorometric Polymerase Activity Assay Kit, Biotium
Supplier: Biotium
29051
89138-880EA 662.91 USD InStock
89138-880
EvaEZ™, Fluorometric Polymerase Activity Assay Kit, Biotium
Assays Enzyme Assays
EvaEZ™ Fluorometric Polymerase Activity Assay Kit provides an easy and accurate way to determine the activity of a nucleic acid polymerase without using radioisotopes.
  • Just add polymerase
  • No radioisotopes needed
  • Can be used at temperatures from 4 °C to 75 °C
EvaEZ™ Fluorometric Polymerase Activity Assay Kit contains EvaGreen® dye together with a primed template, dNTPs, and MgCl2 in a Tris buffer system. In the presence of DNA polymerase activity, the primer will be extended to form a double stranded product that can bind EvaGreen dye, resulting in an increase of fluorescence. The rate of increase of fluorescence is positively correlated to the activity of polymerase. The assay was developed for the measurement of Taq DNA polymerase activity. It can also be used for other DNA polymerases such as Pfu, Vent, Phusion®, Bst, Phi29, MMLV, AMV, SuperScript®, T4 DNA polymerase, T7 DNA polymerase, Klenow, and E. coli DNA polymerase I.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty