Would you like to log in again?
ROX Reference Dye, Biotium
Supplier: Biotium
EvaGreen®
29052
89138-882EA 215.41 USD InStock
89138-882
ROX Reference Dye, Biotium
Assays qPCR Assays
ROX reference dye is used to normalize well-to-well variations that may occur during quantitative, real-time PCR reactions.
  • Red dye used to normalize qPCR reactions
  • Won’t interfere with PCR reactions
  • Supplied as a 25 uM solution
Em/Ex: 600/575 nm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty