Would you like to log in again?
Biotin-20-dUTP, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40030 40030-1
89139-094EA 347.71 USD InStock
89139-094 89139-096
Biotin-20-dUTP, Biotium
Protein Linking and Labeling Systems
Biotin-20-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling. Biotin-dUTP conjugates are available wtih 11-carbon, 16-carbon, or 20-carbon linker lengths.
  • 20-carbon linker between biotin and dUTP
  • Available in solid or solution form
  • Solid contains lyophilized TE buffer
  • Solution is 1 mM in Tris-HCl buffer pH 7.5
In general, the shorter the linker, the more efficiently the biotin-dUTP is incorporated into DNA by DNA polymerases. On the other hand, a longer linker should facilitate interaction between biotin and avidin or streptavidin.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty