Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM in TE buffer (pH 7.5)

Supplier: Biotium


40036
89139-104EA 429.86 USD InStock
89139-104
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM in TE buffer (pH 7.5)
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP)
Biotin-11-dCTP (Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt) can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling. The number 11 is the number of atoms in the linker between biotin and dCTP.


Formula: C₂₈H₄₂N₇O₁₆P₃SLi₄
MW: 885.42 g/mol
Storage Temperature: Freezer
CAS Number: 136632-30-9
Order Now


 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty