Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fura 2-AM ≥98% (by HPLC) fluorescent indicator, for calcium determination

Supplier: Biotium

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


Synonyms: 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl esterFura-2 AM EsterFURA-2 pentakis(acetoxymethyl) esterFURA-2 acetoxymethyl ester

50033 50033-1 50034
89139-210EA 226.82 USD InStock
89139-210 89139-206 89139-208
Fura 2-AM ≥98% (by HPLC) fluorescent indicator, for calcium determination
Fura 2-AM
Fura-2, AM ester is a ratiometric fluorescent dye that binds to free intracellular calcium, and is the preferred dye for ratiometric imaging microscopy. Fura-2, AM ester is membrane-permeant and thus can be loaded into cells via incubation.

  • Kd: 145 nM (22°C in pH 7.2 buffer) (after hydrolysis)
  • λEx/λEm (no Ca2+) = 363/512 nm; λEx/λEm (high Ca2+) = 335/505 nm (after hydrolysis)
  • ε (363 nm, no Ca2+) = 27,000 M-1cm-1; ε (335 nm, high Ca2+) = 35,000 M-1cm-1 (after hydrolysis)
  • Cell permeability: Membrane permeant
  • Light yellow solid soluble in DMSO
  • Store at -20°C and protect from light, especially in solution

Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 (catalog no. 59004) is often used as a dispersing agent to facilitate cell loading. Fura-2, AM ester itself does not bind Ca2+, but it is readily hydrolyzed to Fura-2 by endogenous esterases once it enters cells.

Fura-2 fluorescence can be detected using flow cytometry, fluorescence microscopy, or certain fluoresence microplate readers. Upon calcium binding, the fluorescent excitation maximum of the indicator undergoes a blue shift from 363 nm (Ca2+-free) to 335 nm (Ca2+-saturated), while the fluorescence emission maximum is relatively unchanged at ~510 nm. A major use of Fura-2 is in drug discovery, for measuring intracellular calcium mobilization after activation of G protein-coupled receptors (GPCRs) and calcium channels.


Formula: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036976
CAS Number: 108964-32-5
Order Now


 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty