Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acridine Orange 10-nonyl bromide (NAO) mitochondria dye, for fluorescence microscopy

Supplier: Biotium

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


Warning
Synonyms: Nonyl acridine orange3,6-Bis(dimethylamino)-10-nonylacridinium bromideNAO10-Nonylacridine orange bromide

70012
89139-382EA 147.43 USD InStock
89139-382
Acridine Orange 10-nonyl bromide (NAO) mitochondria dye, for fluorescence microscopy
Acridine Orange 10-nonyl bromide (NAO)
NAO is a green fluorescent mitochondrial dye whose staining is not dependent on mitochondrial membrane potential.

  • Cellular localization: Mitochondria
  • For live or fixed cells: For live/intact cells
  • Assay type/options: No-wash staining, Real-time imaging
  • Potential dependence: Mitochondrial potential-independent
  • Colors: Red
  • Excitation/Emission: 495/522 nm
  • Orange solid soluble in DMSO or DMF
  • Store at 4°C and protect from light, especially in solution

NAO is a green fluorescent mitochondrial dye whose staining is not dependent on mitochondrial membrane potential, unlike JC-1 (70011) or Rhodamine 123 (70010). NAO has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis. The dye is reported to bind cardiolipin, a phosphoprotein in the inner mitochondrial membrane. Also see our MitoView™ Dyes and other mitochondrial dyes.


Formula: C₂₆H₃₈BrN₃
MW: 472.50 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00467605
CAS Number: 75168-11-5
Order Now


 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty