Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biocytin ≥95% (by TLC)

Supplier: Biotium

Synonyms: ε-Biotinoyl-L-lysineH-Lys(biotinyl)-OH

90055
89139-610EA 147.34 USD InStock
89139-610
Biocytin ≥95% (by TLC)
Biocytin
Biocytin (ε-Biotinoyl-L-lysine) is a cellular tracer that can be introduced into cells by microinjection.

  • White solid soluble in water or DMSO

Biocytin is the natural substrate of biotinidase and can be used to measure the biotinidase activity and diagnose biotinidase deficiency in scientific research as a histological stain for nerve cells.


Formula: C₁₆H₂₈N₄O₄S
MW: 372.484 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077319
CAS Number: 576-19-2
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty