Would you like to log in again?
Reactive Biotin Compounds, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
92176 90051 90064-1 92011 90067 90111 90078 90083 92012 92224 90064 92169 90061 90113 90053 90059 90068 90065-1 90069 90080 90050 92167 90054 92168 92010 90063 90112 90066-1 90065 90052 90079 90066 90058
10119-220EA 112.32 USD InStock
10119-220 10119-222 89139-726 89139-804 89139-628 89139-606 89139-608 89139-722 89139-602 89139-668 89139-800 89139-724 89139-802 89139-626 89139-604 89139-630 89139-632 10119-154 89139-638 89139-616 89139-618 89139-634 89139-658 89139-636 89139-642 89139-622 89139-644 89139-600 89139-660 89139-640 89139-662 10119-224 89411-598
Reactive Biotin Compounds, Biotium
Protein Expression Protein Expression Reagents
Biotinylation reagents with a variety of reactive functional groups.
  • Reagents for biotinylation of proteins and other biomolecules
  • Full selection of reactive functional groups
  • Variety of linker lengths available for improved water solubility and streptavidin binding
Biotium offers reactive biotin compounds with a variety of functional groups:
Amine reactive:
Succinimidyl esters react with primary and secondary amines and can be used to label proteins or peptides
The Biotin SE Protein Labeling Kit includes everything you need to label and purify three reactions of up to 1 mg protein each with biotin SE
Thiol reactive:
Maleimide and iodoacetamide react with thiol groups
MTSEA selectively labels thiols under mild conditions
TS biotin compounds allow selective labeling of cell surface thiols
Other reactive chemistries:
Amine groups can be reacted with activated carboxylic acids
Cadaverine can be coupled to carboxylic acids, DNA, and other biomolecules
Cadaverine free base can be directly coupled to activated carboxylic acids without the use of additional base
Hydrazides react with aldehydes or ketones to form unstable hydrazone linkages
Aminooxy reacts with aldehydes or ketones to form stable oxime bonds, and may be preferable to hydrazides for labeling
Tyramides can be used for HRP-mediated tyramide signal amplification (TSA) reactions
Alkynes react with azides by copper(I)-catalyzed 1,3-dipolar Huisgen cycloaddition
BCN groups react with azides by copper-free 1,3-dipolar Huisgen cycloaddition
Also see our selection of fluorescent CF™ Dye Streptavidin and Monoclonal Mouse Anti-Biotin Conjugates.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty