Would you like to log in again?
Reactive Dye Conjugates, Biotium
Supplier: Biotium
90110 92000 90108 90104 90107 90116 90106 90073 90115 90103 92001 90109 90105
89139-708EA 240.51 USD InStock
89139-708 89139-716 89139-706 89139-718 89139-712 89139-714 89139-730 89139-796 89139-720 89139-732 89139-710 89139-650 89139-794
Reactive Dye Conjugates, Biotium
Protein Linking and Labeling Systems
Reactive dyes containing an amine functional group.
  • Aminooxy conjugates
  • Cadaverine conjugates
  • Can be used to label abasic sites in damaged DNA
Our aminooxy reagents such as aminooxy-5(6)-FAM react with aldehydes or ketones to form a stable hydrazone compound under mild conditions
Dye conjugates to PEO3-amine have a long water-soluble PEG spacer that separates the fluorescent dye and the amine functional group. These dyes can be coupled to activated carboxy groups or sulfonyl chlorides.
Cadaverine conjugates can be coupled to activated carboxy groups or sulfonyl chlorides.
Similar to ARP, aminooxy-5(6)-FAM can be used to label abasic sites in damaged DNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty