Would you like to log in again?
Thiol-Reactive Fluorescent Dyes and Other Thiol-Modifying Reagents, Biotium
Supplier: Biotium
91046 91053 91031 91038 91055 91044 91022 91010 91023 91032 91025 91043 91045 92015 91030 92013 91048 91028 92016 91024 91037 91052 91027 91029 91042 91011 91026 91036 91035 91021 91033
89139-748EA 99.69 USD InStock
89139-748 89139-806 89139-788 89139-744 89139-766 89139-746 89139-768 89139-752 89139-754 89139-776 89493-048 89139-770 89139-750 89410-782 89139-738 89493-050 89139-810 89139-734 89139-778 89139-756 89410-784 89139-812 89139-758 89139-784 89139-740 89139-762 89139-742 89139-764 89139-780 89139-760 89139-782
Thiol-Reactive Fluorescent Dyes and Other Thiol-Modifying Reagents, Biotium
Protein Linking and Labeling Systems
MTS, TS, and maleimide, and other thiol-modifying reagents and reactive fluorescent dyes.
  • Selection of thiol-reactive fluorescent dyes in a variety of colors
  • Thiol-reactive cross-linkers and other modifiers
  • Available with MTS, TS, and maleimide reactive groups
  • Other thiol-reactive probes for specialized applications available
Biotium offers a variety of methanethiosulfonate (MTS) thiol modifying reagents and cross-linkers, as well as charged thiosulfate (TS) reagents for selectively modifying cell surface thiols.
In addition, we offer thiol-reactive probes for specialized applications. Eosin-5-maleimide is an thiol-reactive photosensitizer that can be used for photoconversion of electron-rich compounds like DAB or fluorescent dyes into electron-dense materials for high-resolution electron microscopy studies. It is also a good fluorescence acceptor for fluorescein, dansyl, and coumarin dyes for fluorescence resonance energy transfer (FRET). CPM (7-diethylamino-3-(4′-maleimidylphenyl)-4-methylcoumarin) is probably the most widely used blue fluorescent thiol-reactive dye. This maleimide derivative of coumarin is essentially nonfluorescent until it reacts with thiols, making it possible to quantify thiols without a separation step. ABD-F is a useful reagent for the sensitive and specific detection of thiols both by TLC and HPLC. When combined with a reducing agent such as tributylphosphine, it can also be used for the determination of disulfides in peptides and proteins. INA (5-iodonaphthyl-1-azide) is a lipophilic photoreactive probe, which has been used to selectively label membrane-embedded cysteine residues of proteins. N-(1,10-Phenanthrolin-5-yl)iodoacetamide has been used to label thiols to confer the metal-binding properties of the phenanthroline chelator. For example, completing of the chelator with copper can be used to selectively cleave RNA or DNA in the presence of hydrogen peroxide.
See Reactive Biotin Compounds for thiol-reactive biotinylation reagents. Also see our line of next-generation fluorescent CF™ Dyes, available in a wide selection of reactive dye formats, including thiol-reactive maleimide and MTS forms.
Texas Red is a registered trademark of Thermo Fisher Scientific.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty