Would you like to log in again?
VivoBrite™ Rapid Antibody Labeling Kits for Small Animal In Vivo Imaging, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
92160 92163 92162 92161
89411-570EA 554.5 USD InStock
89411-570 89139-850 89139-848 89139-846
VivoBrite™ Rapid Antibody Labeling Kits for Small Animal In Vivo Imaging, Biotium
Protein Linking and Labeling Systems
Near-infrared dyes offer highly sensitive and specific detection in complex biological systems, because cells and tissues have intrinsically low autofluorescence in the near-IR region
VivoBrite Antibody Labeling Kits include amine-reactive near-IR CF™ dye and all other components required for antibody labeling, purification, and filter sterilization for small animal injection. Available with your choice of CF™680, CF™750, CF™770, or CF™790 dyes.
The long emission wavelengths of near-infrared fluroescent dyes also have strong tissue penetration, permitting in vivo fluorescence imaging in small animals. Near-infrared dyes tend to suffer from limited fluorescence brightness to dye aggregation and poor stability. Biotium's next-generation near-IR dyes are designed to overcome these problems, resulting in extremely bright and photostable near-IR fluorescence.
Ordering information: Each kit is sufficient for three labelings of 1 mg IgG per reaction.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty