Would you like to log in again?
Neptune® S³ Low Retention Pipette Tips
Supplier: Biotix
Neptune
2372 2090 63300544 63302167 63302177 2102.S 2100 2092 63302158 2102 2100.Y 2101 63302157 2372.S 2092.S 63300508
89168-734CS 295.42 USD InStock
89168-734 89168-758 89168-736 89168-732 89168-760 89140-098 89140-096 89140-130 89140-884 89425-648 89140-100 89140-892 89140-128 89140-898 89140-106 89140-888
Neptune® S³ Low Retention Pipette Tips
Pipette Tips
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips are made from virgin polypropylene and are equipped with a thin wall design that provides an optimal seal with industry leading pipettors.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Ordering information: A variety of packaging options are available. Bulk orders are packaged 1000 per bag. Rack and Stack packaging contains five inserts of 192 tips. Standard rack packaging contains 96 tips per rack.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty