Would you like to log in again?
VWR® Traceable® Flashing LED Alert Big-Digit Dual Channel Timer
Supplier: VWR
VWR®
90009-77
89140-196EA 51.68 USD InStock
89140-196
VWR® Traceable® Flashing LED Alert Big-Digit Dual Channel Timer
Timers Digital Timers
Allows simultaneous programming of two different activities from 99 hours, 59 minutes, 59 seconds to 1 second.
  • Bright LED digits
  • Memory function for repetitive timing
  • Unique visual indicator during countdown
Resolution is 1 second; accuracy is 0.01%
For repetitive timing, the microcomputer chip permits setting 2 countdown times into memory. The memory recalls the last countdown time set. Large ⁵/₈-inch high LCD digits may be read from 18 feet.
Unique visual indicator for each channel when counting down, green light flashes every second once the countdown time reaches 10 seconds. Red light flashes when zero has been reached and the alarm is sounding. Alarm has three positions which allow setting to high decibel for noisy areas, low decibel for quiet environments, or off the muted setting for silent, visual flashing LED-only operation.
Each channel has a distinct electronic alarm and visual indicator when zero is reached. Audible alarms turns off automatically after one minute or may be silenced automatically.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with magnetic back, stand/spring-clip, opening for a lanyard, two alkaline batteries and Traceable® Certificate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty