Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CLINITEK Advantus® Urine Chemistry Analyzer, Siemens Healthineers

Supplier: Siemens Healthineers
CLINITEK® CLINITEK Status®
STARTAD 1420
76313-768EA 11364.36 USD InStock
76313-768 89140-206
CLINITEK Advantus® Urine Chemistry Analyzer, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Analysers
CLINITEK® Advantus® Urine Chemistry Analyzer from Siemens Healthcare Diagnostics provides automated reading of the Multistix® family of urinalysis tests.

  • Easy-to-use color touchscreen
  • Consolidate microscopy results onto one patient record
  • Automatic QC prompting and QC lockout capability
  • Three sieve functions
  • Convenient data retrieval
  • Improved data integrity

The CLINITEK Advantus® Urine Chemistry Analyzer allows labs to respond to demands for higher productivity and high quality

A total of three sieve functions; confirmatory and microscopic to flag abnormal results for quick review. Convenient data retrieval for up to 500 patients by patient ID, and 200 control results. Improved data integrity by automatically identifying and reporting results only from validated Siemens strip types. Improved workflow eliminates the need for manually selecting urinalysis strip type. Throughput of 7 seconds/sample; up to 500 tests/hour.

Optional barcode scanner available separately.

Streamline your workflow with flexible operation. Immediate start-up. Automatic calibration. Network ready. A wide range of options. Added QC features. Flexible operation to meet your needs. Auto-checks for strip identification and humidity exposure improves data integrity, workflow and helps ensure quality results. Consistent, reliable results, with one-touch switching between test strip configurations. Automatic prompts and test lock-out if QC is not run for improved quality control. Automatically read, interpreted, and documented Multistix® test strip results.

Additional information
Dimensions: 12.75 (h) x 15.75 (w) x 13.75 (d) in. / 32 (h) x 39 (w) x 35 (d) cm
Weight: 7.2 kg (16 lbs)
Power Requirements: 100 V to 240 V; 50/60 Hz

Certifications: UL listed, CE marked, EMC compliant.

Ordering information: Optional barcode scanner available separately.

Order Now
 

ORDER

ACCESSORIES

COMPLEMENTARY PRODUCTS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty