Would you like to log in again?
CLINITEK Status®+ Urine Analyzer, Siemens Healthineers
Supplier: Siemens Healthineers
CLINITEK®
STARTUA 1780
89140-210EA 1941.36 USD InStock
89140-210 76313-712
CLINITEK Status®+ Urine Analyzer, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Analysers
CLINITEK Status®+ Urine Analyzer from Siemens Healthineers features automatic checks (Auto-Checks)
  • Auto-Checks for enhanced confidence
  • Improved clinical information
  • For routine testing and detection
  • Ready for connectivity to DMS
The analyzer automatically checks each test strip for humidity exposure, common sample interferences, and strip identification to identify Siemens test strips. These Auto-Checks provide improved clinical information and enhance the confidence in clinical decisions when performed on every test. This also ensures consistent results interpretation with automatic timing and reading of every urine specimen. The CLINITEK Status®+ Urine Analyzer offers a comprehensive point-of-care urinalysis menu for routine testing, detection of kidney disease, and hCG pregnancy testing.
The CLINITEK Status®+ Urine Analyzer is ready for connectivity to data management solutions (DMS) via a simple upgrade kit (see CLINITEK Status Connect System).
Order Now
ORDER
ACCESSORIES
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty