Would you like to log in again?
GLOVE CUT RESISTANT JUMBO
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Cut Resistant Gloves Perfect Fit Jumbo/13-Cut^ Color Black.ANSI/ISEA Cut Level 2. ANSI/ISEA Abrasion Level 1. Application: Material Handling, Cut Resistant Glove, Seamless Knit, Size XL, Material Kevlar(R), Gauge Lightweight
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE CUT RESISTANT JUMBO 89140-248 KV13AJ Case of 288 Retrieving
Pack of 24 Retrieving
Pair Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty