Would you like to log in again?
VWR® Rota-Rack® Duo Tube Rack
Supplier: VWR
VWR®
HS29041AV
89140-272CS 3713.72 USD InStock
89140-272
VWR® Rota-Rack® Duo Tube Rack
Racks Tube Racks
Dilute or take off concentrate using one rack with the use of the Duo Tube Rack.
  • Different orientations provide unique dilution options
  • Tapered wells securely suspend tubes upright
  • Easy removal of tubes
  • Autoclavable
  • Quickly identify tubes with imprinted alphanumeric system
The 0.2 mL tube wells are oriented in standard 96-well format. Use with PCR transfer trays. Rotate rack to customize applications. Rotate to work with ninety-six 0.2 mL tubes and forty-eight 0.5 mL tubes. Turn a module and work with ninety-six 0.2 mL tubes and forty-eight 1.5 mL tubes. Flip rack and work with forty-eight 0.5 mL tubes and forty-eight 1.5 mL tubes. Or turn a module and work with a total of ninety-six 1.5 mL tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty