Would you like to log in again?
VWR® Sealing Film for Automation
Supplier: VWR
VWR®
RFNL80X100 RRTSNL79X65 RBNL81X100
89140-338EA 753.98 USD InStock
89140-338 89140-336 89140-340
VWR® Sealing Film for Automation
Seals Microplate Seals
Adhesive sealing film or foil rolls constructed on plastic cores, for use with high throughput automated microplate sealers.
  • Continous rolls - no splices
  • Film or foil does not extend beyond edge of plastic core
  • Robust static-free packaging provides protection during shipment and facilitates accurate alignment and adhesion
Sealing film for qPCR, sitting-drop protein crystallisation, HTS and cold storage
Clear polyolefin with ultra-strong, pressure-sensitive silicone adhesive for qPCR, sitting-drop protein crystallisation, HTS, storage and general incubation applications.
- DMSO resistant
- Nuclease-, nucleic acid- and pyrogen-free
- Provides as strong a bond to the plate as heat sealing films without the drawbacks of heat
Sealing foil for PCR, HTS and cold storage
Pierceable aluminium foil with pressure-sensitive acrylic adhesive for PCR, HTS and cold storage.
- DMSO resistant
- Easily pierceable for sample recovery
- Nuclease-, nucleic acid- and pyrogen-free
Sealing film for cell and tissue culture
Breathable rayon films for cell and tissue culture.
- Non cytotoxic
- Highly gas permeable
- Easily pierceable for sample recovery
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty