Would you like to log in again?
VWR THERMOMETER BP EC -50/300C
Supplier: VWR International
VWR Economy Digital Thermometer^ Bottle Probe. Temperature Range 32 to 572°F and 0 to 300°C, Resolution 0.1. Dual Display. Hi/Lo Alarm. Min/Max Memory. Bottle Probe 59 Inch Cable. Countdown Timer. On/Off And Hold Function. C/F Selectable.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR THERMOMETER BP EC -50/300C 89140-490 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty